CH6371379363 Chem Impex International, Inc.

D-Leu-Gly-OH

  • MDL number MFCD00067198
  • Cas Number : 997-05-7
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-leu-gly-oh/3919?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-leu-gly-oh/3919?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart