CH6371379364 Chem Impex International, Inc.

D-Leucyl-leucine

  • MDL number MFCD00069976
  • Cas Number : 38689-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Leu-Leu-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-leucyl-leucine/3920?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-leucyl-leucine/3920?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart