CH6371379366 Chem Impex International, Inc.

Ne-Acetyl-D-lysine

  • MDL number MFCD00237141
  • Cas Number : 51621-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Lys(Ac)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-e-sup-acetyl-d-lysine/3924?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-e-sup-acetyl-d-lysine/3924?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart