CH6371379368 Chem Impex International, Inc.

Ne-Z-D-lysine benzyl ester hydrochloride

  • MDL number MFCD00065241
  • Cas Number : 156917-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Lys(Z)-OBzl HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-z-d-lysine-benzyl-ester-hydrochloride/3926?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-z-d-lysine-benzyl-ester-hydrochloride/3926?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart