CH6371379369 Chem Impex International, Inc.

Nd-Z-D-ornithine

  • MDL number MFCD00063115
  • Cas Number : 16937-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Orn(Z)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-d-sup-z-d-ornithine/3927?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-d-sup-z-d-ornithine/3927?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart