CH6371379375 Chem Impex International, Inc.

D-Proline tert-butyl ester

  • MDL number MFCD00083683
  • Cas Number : 90071-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-proline-i-tert-i-butyl-ester/3934?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-proline-i-tert-i-butyl-ester/3934?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart