CH6371379376 Chem Impex International, Inc.

O-Acetyl-D-serine hydrochloride

  • MDL number MFCD00672365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-162
Synonym D-Ser(Ac)-OH HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-acetyl-d-serine-hydrochloride/3935?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-acetyl-d-serine-hydrochloride/3935?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart