CH6371379377 Chem Impex International, Inc.

O-Benzyl-D-threonine amide hydrochloride

  • MDL number MFCD00237280
  • Cas Number : 201275-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Thr(Bzl)-NH2 HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-d-threonine-amide-hydrochloride/3937?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-benzyl-d-threonine-amide-hydrochloride/3937?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart