CH6371379381 Chem Impex International, Inc.

O-Methyl-D-tyrosine

  • MDL number MFCD00190961
  • Cas Number : 39878-65-4
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-methyl-d-tyrosine/3942?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-methyl-d-tyrosine/3942?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart