CH6371379382 Chem Impex International, Inc.

D-Valine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00136545
  • Cas Number : 17662-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Val-OBzl TosOH
Storage temp. Store at 0-8
Physical Form Crystalline
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-valine-benzyl-ester-4-toluenesulfonate-salt/3943?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-valine-benzyl-ester-4-toluenesulfonate-salt/3943?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart