CH6371379393 Chem Impex International, Inc.

DL-Aspartic acid b-methyl ester hydrochloride

  • MDL number MFCD00214167
  • Cas Number : 1835-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym DL-Asp(OMe)-OH�
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-aspartic-acid-b-methyl-ester-hydrochloride/3956?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-aspartic-acid-b-methyl-ester-hydrochloride/3956?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart