CH6371379398 Chem Impex International, Inc.

L-Glutamic acid gamma-anilide

  • MDL number MFCD00063105
  • Cas Number : 5963-60-0
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-anilide/3963?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-anilide/3963?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart