CH6371379405 Chem Impex International, Inc.

Glu-Asp-OH

  • MDL number MFCD00037308
  • Cas Number : 3918-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-alpha-Glutamyl-L-aspartic acid
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glu-asp-oh/3970?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glu-asp-oh/3970?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart