CH6371379406 Chem Impex International, Inc.

Glu-Gly-OH

  • MDL number MFCD00037311
  • Cas Number : 13716-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-alpha-Glutamyl-glycine
Purity 0.99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glu-gly-oh/3971?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glu-gly-oh/3971?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart