CH6371379407 Chem Impex International, Inc.

L-Glutamic acid alpha-4-nitroanilide

  • MDL number MFCD00038111
  • Cas Number : 24032-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-187 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white to yellowish crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-4-nitroanilide/3972?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-4-nitroanilide/3972?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart