CH6371379410 Chem Impex International, Inc.

Glycine amide acetate

  • MDL number MFCD00237951
  • Cas Number : 105359-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-125 �C
Synonym Gly-NH2 AcOH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glycine-amide-acetate/3977?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glycine-amide-acetate/3977?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart