CH6371379413 Chem Impex International, Inc.

Nim-Benzyl-L-histidine

  • MDL number MFCD00038630
  • Cas Number : 16832-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-His(Bzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-benzyl-l-histidine/3980?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-benzyl-l-histidine/3980?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart