CH6371379414 Chem Impex International, Inc.

Nim-Benzyl-L-histidine methyl ester dihydrochloride

  • MDL number MFCD00237097
  • Cas Number : 93983-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-benzyl-l-histidine-methyl-ester-dihydrochloride/3981?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-benzyl-l-histidine-methyl-ester-dihydrochloride/3981?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart