CH6371379415 Chem Impex International, Inc.

Nim-4-Methyltrityl-L-histidine

  • MDL number MFCD00237169
  • Cas Number : 133367-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-216
Synonym L-His(Mtt)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-4-methyltrityl-l-histidine/3982?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-4-methyltrityl-l-histidine/3982?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart