CH6371379417 Chem Impex International, Inc.

Nim-Trityl-L-histidine tert-butyl ester hydrochloride

  • MDL number MFCD00798683

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-sup-im-sup-trityl-l-histidine-i-tert-i-butyl-ester-hydrochloride/3984?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-sup-im-sup-trityl-l-histidine-i-tert-i-butyl-ester-hydrochloride/3984?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart