CH6371379419 Chem Impex International, Inc.

His-Gly-OH

  • MDL number MFCD00037863
  • Cas Number : 2578-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/his-gly-oh/3986?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/his-gly-oh/3986?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart