CH6371379421 Chem Impex International, Inc.

L-Histidine b-naphthylamide

  • MDL number MFCD00051166
  • Cas Number : 7424-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-His-betaNA
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-histidine-ß-naphthylamide/3989?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-histidine-ß-naphthylamide/3989?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart