CH6371379422 Chem Impex International, Inc.

His-Val-OH

  • MDL number MFCD00037856
  • Cas Number : 76019-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Off white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/his-val-oh/3990?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/his-val-oh/3990?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart