CH6371379427 Chem Impex International, Inc.

trans-L-4-Hydroxyproline b-naphthylamide hydrochloride

  • MDL number MFCD00237996
  • Cas Number : 201994-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Hyp-betaNA HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-trans-i-l-4-hydroxyproline-ß-naphthylamide-hydrochloride/3996?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-trans-i-l-4-hydroxyproline-ß-naphthylamide-hydrochloride/3996?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart