CH6371379429 Chem Impex International, Inc.

Ile-Ile-OH

  • MDL number MFCD00038574
  • Cas Number : 42537-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258-262
Synonym L-Isoleucyl-L-isoleucine
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ile-ile-oh/4000?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ile-ile-oh/4000?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart