CH6371379431 Chem Impex International, Inc.

L-Isoleucine 4-nitroanilide

  • MDL number MFCD00238006
  • Cas Number : 70324-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-62
Synonym L-Ile-pNA
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow to white powder with tubbers
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-isoleucine-4-nitroanilide/4003?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-isoleucine-4-nitroanilide/4003?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart