CH6371379433 Chem Impex International, Inc.

L-Leucine a-naphthylamide

  • MDL number MFCD00083643
  • Cas Number : 203793-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-85
Synonym L-Leu-alphaNA
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-leucine-alpha-naphthylamide/4005?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-leucine-alpha-naphthylamide/4005?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart