CH6371379434 Chem Impex International, Inc.

L-Leucine 7-amido-4-methylcoumarin

  • MDL number MFCD00056095
  • Cas Number : 66447-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-171 °C
Purity 99.5
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-leucine-7-amido-4-methylcoumarin/4006?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-leucine-7-amido-4-methylcoumarin/4006?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart