CH6371379438 Chem Impex International, Inc.

Ne-Boc-L-lysine allyl ester hydrochloride

  • MDL number MFCD00798677
  • Cas Number : 218938-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Lys(Boc)-OAll HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-l-lysine-allyl-ester-hydrochloride/4013?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-l-lysine-allyl-ester-hydrochloride/4013?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart