CH6371379439 Chem Impex International, Inc.

Ne-Boc-L-lysine 4-nitroanilide

  • MDL number MFCD00237144
  • Cas Number : 172422-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-97
Synonym L-Lys(Boc)-pNA
Purity 99
Storage temp. Store at 0-8
Physical Form White to light yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-l-lysine-4-nitroanilide/4014?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-l-lysine-4-nitroanilide/4014?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart