CH6371379440 Chem Impex International, Inc.

Ne-Fmoc-L-lysine

  • MDL number MFCD00038365
  • Cas Number : 84624-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Lys(Fmoc)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-fmoc-l-lysine/4015?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-fmoc-l-lysine/4015?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart