CH6371379443 Chem Impex International, Inc.

3-Fluoro-D-phenylalanine methyl ester hydrochloride

  • MDL number MFCD00672371
  • Cas Number : 201479-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-159 �C
Synonym D-Phe(3-F)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-fluoro-d-phenylalanine-methyl-ester-hydrochloride/4019?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-fluoro-d-phenylalanine-methyl-ester-hydrochloride/4019?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart