CH6371379448 Chem Impex International, Inc.

Nd-Fmoc-L-ornithine

  • MDL number MFCD00237208
  • Cas Number : 147071-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-146
Synonym L-Orn(Fmoc)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-fmoc-l-ornithine/4025?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-fmoc-l-ornithine/4025?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart