CH6371379449 Chem Impex International, Inc.

Nd-Pyrazinylcarbonyl-L-ornithine

  • MDL number MFCD00672359
  • Cas Number : 201047-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238-244 �dec.)
Synonym L-Orn(pyrazinylcarbonyl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-pyrazinylcarbonyl-l-ornithine/4026?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-pyrazinylcarbonyl-l-ornithine/4026?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart