CH6371379450 Chem Impex International, Inc.

Ndelta-Z-L-ornithine benzyl ester hydrochloride

  • MDL number MFCD00237210
  • Cas Number : 63594-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-162 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-z-l-ornithine-benzyl-ester-hydrochloride/4027?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-delta-sup-z-l-ornithine-benzyl-ester-hydrochloride/4027?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart