CH6371379455 Chem Impex International, Inc.

4-Amino-D-phenylalanine hydrochloride

  • MDL number MFCD00236783
  • Cas Number : 126257-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-amino-d-phenylalanine-hydrochloride/4034?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-amino-d-phenylalanine-hydrochloride/4034?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart