CH6371379456 Chem Impex International, Inc.

4-Amino-DL-phenylalanine

  • MDL number MFCD00007917
  • Cas Number : 2922-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-amino-dl-phenylalanine/4035?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-amino-dl-phenylalanine/4035?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart