CH6371379459 Chem Impex International, Inc.

4-Chloro-D-phenylalanine methyl ester hydrochloride

  • MDL number MFCD00236920
  • Cas Number : 33965-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Phe(4-Cl)-OMe HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-chloro-d-phenylalanine-methyl-ester-hydrochloride/4038?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-chloro-d-phenylalanine-methyl-ester-hydrochloride/4038?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart