CH6371379463 Chem Impex International, Inc.

O-Methyl-DL-tyrosine

  • MDL number MFCD00063071
  • Cas Number : 7635-29-2
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-methyl-dl-tyrosine/4042?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-methyl-dl-tyrosine/4042?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart