CH6371379465 Chem Impex International, Inc.

S-Trityl-L-penicillanine

  • MDL number MFCD00237408
  • Cas Number : 135681-66-2
  • Molecular Weight : 391.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-trityl-l-penicillanine/4044?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-trityl-l-penicillanine/4044?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart