CH6371379466 Chem Impex International, Inc.

L-Phenylalanine chloromethylketone hydrochloride

  • MDL number MFCD00133443
  • Cas Number : 52735-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Phenylalanine chloromethylketone hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form Whtie solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-phenylalanine-chloromethylketone-hydrochloride/4046?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-phenylalanine-chloromethylketone-hydrochloride/4046?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart