CH6371379470 Chem Impex International, Inc.

L-Phenylalanine pyrrolidide

  • MDL number MFCD00672358
  • Cas Number : 56414-89-2
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 78 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-phenylalanine-pyrrolidide/4050?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-phenylalanine-pyrrolidide/4050?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart