CH6371379474 Chem Impex International, Inc.

L-Proline 4-methoxy-b-naphthylamide hydrochloride

  • MDL number MFCD00058032
  • Cas Number : 100930-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Pro-4MbetaNA HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-proline-4-methoxy-beta-naphthylamide-hydrochloride/4055?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-proline-4-methoxy-beta-naphthylamide-hydrochloride/4055?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart