CH6371379476 Chem Impex International, Inc.

L-Proline ethylamide hydrochloride

  • MDL number MFCD00238159
  • Cas Number : 58107-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-124 �C
Synonym L-Pro-NHEt HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-proline-ethylamide-hydrochloride/4058?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-proline-ethylamide-hydrochloride/4058?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart