CH6371379479 Chem Impex International, Inc.

L-Proline 4-nitroanilide hydrobromide

  • MDL number MFCD00672477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Pro-pNA HBr
Purity 99
Storage temp. Store at 0-8
Physical Form Light beige crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-proline-4-nitroanilide-hydrobromide/4062?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-proline-4-nitroanilide-hydrobromide/4062?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart