CH6371379480 Chem Impex International, Inc.

O-tert-Butyl-L-serine t-butyl ester

  • MDL number MFCD00080900
  • Cas Number : 48067-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ser(tBu)-OtBu
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-serine-t-butyl-ester/4064?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-serine-t-butyl-ester/4064?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart