CH6371379488 Chem Impex International, Inc.

b-Ala-b-Ala-OH

  • MDL number MFCD00038199
  • Cas Number : 2140-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(3-Amino-propionylamino)propionic acid
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-ala-beta-ala-oh/4077?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-ala-beta-ala-oh/4077?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart