CH6371379489 Chem Impex International, Inc.

beta-Alanine beta-naphthylamide hydrobromide

  • MDL number MFCD00080853
  • Cas Number : 201985-01-5
  • Molecular Weight : 295.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form Light beige powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-alanine-ß-naphthylamide-hydrobromide/4078?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-alanine-ß-naphthylamide-hydrobromide/4078?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart