CH6371379490 Chem Impex International, Inc.

b-Chloro-L-alanine

  • MDL number MFCD00046536
  • Cas Number : 2731-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym beta-Chloro-L-Ala-OH
Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-chloro-l-alanine/4080?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-chloro-l-alanine/4080?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart