CH6371379494 Chem Impex International, Inc.

beta-Chloro-DL-alanine hydrochloride

  • MDL number MFCD00066125
  • Cas Number : 35401-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 176 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/beta-chloro-dl-alanine-hydrochloride/4084?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/beta-chloro-dl-alanine-hydrochloride/4084?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart